siRNA Details
| Gene Symbol: Palmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023245.3 | NP_075734.3 | 4 | 745 | ||
| GenBank mRNA | AF263246.1 | - | 4 | 449 | ||
| AJ312215.1 | - | 4 | 468 | |||
| AK014531.1 | - | 4 | 543 | |||
| AK136939.1 | - | 4 | 652 | |||
| AK171869.1 | - | 4 | 476 | |||
| AL391037.1 | - | 4 | 449 | |||
| BC010193.1 | - | 4 | 509 | 3156945 | ||

Palmd
Genomic Map