siRNA Details
| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038643.1 | NP_001033732.1 | 3 | 857 | ||
| NM_023908.2 | NP_076397.2 | 3 | 857 | |||
| GenBank mRNA | AF226324.1 | - | 3 | 755 | ||
| AK037938.1 | - | 3 | 672 | |||
| AK046945.1 | - | 2 | 217 | |||
| AK151781.1 | - | 3 | 827 | |||
| AK152488.1 | - | 3 | 691 | |||
| AK166266.1 | - | 3 | 628 | |||
| BC025059.1 | - | 3 | 690 | 2598913 | ||
| BC094464.1 | - | 3 | 649 | |||

Slco3a1
Genomic Map