siRNA Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024187.4 | NP_077149.2 | 2, 3 | 188 | ||
| GenBank mRNA | AK008332.1 | - | 2, 3 | 150 | ||
| AK012334.1 | - | 2, 3 | 148 | |||
| AK012849.1 | - | 2, 3 | 148 | |||
| AK088979.1 | - | 2, 3 | 158 | |||
| AK150872.1 | - | 2, 3 | 188 | |||
| BC002184.1 | - | 2, 3 | 127 | |||
| BC069888.1 | - | 2, 3 | 130 | |||
| BC115479.1 | - | 2, 3 | 150 | 40100558 | ||
| BC115480.1 | - | 2, 3 | 150 | 40100559 | ||

U2af1
Genomic Map