siRNA Details
Gene Symbol: Jkamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205067.1 | NP_001191996.1 | 4 | 645 | ||
NM_024205.2 | NP_077167.1 | 4 | 663 | |||
GenBank mRNA | AK004555.1 | - | 4 | 600 | ||
BC002238.1 | - | 4 | 379 | 3491879 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
