siRNA Details
Gene Symbol: Ttc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267622.1 | NP_001254551.1 | 23 | 7837 | ||
GenBank mRNA | AK031458.1 | - | 6 | 2431 | ||
AK081382.1 | - | 6 | 2309 | |||
BC046779.1 | - | 17 | 5501 | 5719497 | ||
BC060735.1 | - | 16 | 4497 |
