siRNA Details
| Gene Symbol: Cox6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025628.2 | NP_079904.1 | 4 | 438 | ||
| GenBank mRNA | AK007996.1 | - | 4 | 439 | ||
| AK011089.1 | - | 3 | 403 | |||
| AK012686.1 | - | 4 | 405 | |||
| AK018721.1 | - | 4 | 373 | |||
| BC024343.1 | - | 4 | 381 | 4989436 | ||

Cox6b1
Genomic Map