siRNA Details
| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025898.3 | NP_080174.1 | 2 | 163 | ||
| GenBank mRNA | AK004826.1 | - | 2 | 172 | ||
| AK005372.1 | - | 2 | 165 | |||
| AK013895.1 | - | 2 | 182 | |||
| AK050623.1 | - | 2 | 227 | |||
| AK136689.1 | - | 2 | 166 | |||
| AK171998.1 | - | 2 | 227 | |||
| BC004804.1 | - | 2 | 194 | 3584869 | ||

Napa
Genomic Map