siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025900.2 | NP_080176.2 | 10, 11 | 1297 | ||
| GenBank mRNA | AK007546.1 | - | 7, 8 | 846 | ||
| AK033451.1 | - | 10, 11 | 1187 | |||
| AK141790.1 | - | 4, 5 | 649 | |||
| BC055451.1 | - | 10, 11 | 1243 | 4164972 | ||

Dek
Genomic Map