siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025900.2 | NP_080176.2 | 11 | 1313 | ||
| GenBank mRNA | AK007546.1 | - | 8 | 862 | ||
| AK033451.1 | - | 11 | 1203 | |||
| AK141790.1 | - | 5 | 665 | |||
| BC055451.1 | - | 11 | 1259 | 4164972 | ||

Dek
Genomic Map