siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025900.2 | NP_080176.2 | 11 | 1385 | ||
| GenBank mRNA | AK007546.1 | - | 8 | 934 | ||
| AK033451.1 | - | 11 | 1275 | |||
| AK141790.1 | - | 5 | 737 | |||
| BC055451.1 | - | 11 | 1331 | 4164972 | ||

Dek
Genomic Map