siRNA Details
| Gene Symbol: Cts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026906.3 | NP_081182.2 | 5 | 670 | ||
| NM_054092.2 | NP_473433.2 | 5 | 595 | |||
| GenBank mRNA | AF456464.1 | - | 4 | 455 | ||
| AK005389.1 | - | 5 | 593 | |||
| AY034574.1 | - | 5 | 609 | |||
| BC099388.1 | - | 5 | 564 | 30786828 | ||

Cts3
Genomic Map