siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 3, 4 | 396 | ||
| NM_027307.4 | NP_081583.3 | 4, 5 | 532 | |||
| GenBank mRNA | AK009072.1 | - | 2, 3 | 203 | ||
| AK133865.1 | - | 3, 4 | 398 | |||
| AK167646.1 | - | 3, 4 | 373 | |||
| BC011152.1 | - | 3, 4 | 357 | 4188434 | ||
| BC098486.1 | - | 4, 5 | 528 | 30024953 | ||
| CD552063.1 | - | 4, 5 | 493 | |||
| CK391733.1 | - | 3, 4 | 396 | |||

Golm1
Genomic Map