siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 9 | 1072 | ||
| NM_027307.4 | NP_081583.3 | 10 | 1208 | |||
| GenBank mRNA | AK009072.1 | - | 8 | 879 | ||
| AK133865.1 | - | 9 | 1074 | |||
| BC011152.1 | - | 9 | 1033 | 4188434 | ||
| BC098486.1 | - | 10 | 1204 | 30024953 | ||

Golm1
Genomic Map