siRNA Details
| Gene Symbol: Fam110c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027828.2 | NP_082104.2 | 1 | 919 | ||
| GenBank mRNA | AK018545.1 | - | 1 | 894 | ||
| AK139763.1 | - | 1 | 1027 | |||
| AK160079.1 | - | 1 | 923 | |||
| BC019466.1 | - | 1 | 919 | 4458596 | ||
| BC085288.1 | - | 1 | 904 | 5354365 | ||

Fam110c
Genomic Map