siRNA Details
| Gene Symbol: Ms4a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025610.4 | NP_001020781.1 | 4 | 502 | ||
| NM_001276398.1 | NP_001263327.1 | 5 | 799 | |||
| NM_027836.6 | NP_082112.3 | 5 | 559 | |||
| GenBank mRNA | AF237917.2 | - | 5 | 510 | ||
| AK018686.1 | - | 4 | 473 | |||
| AK046559.1 | - | 5 | 560 | |||
| AK079888.1 | - | 5 | 800 | |||
| AK149890.1 | - | 5 | 527 | |||
| AK151261.1 | - | 5 | 530 | |||
| AK152098.1 | - | 5 | 530 | |||
| BC019784.1 | - | 4 | 449 | |||
| BC024402.1 | - | 5 | 518 | 5035752 | ||

Ms4a7
Genomic Map