siRNA Details
| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028083.4 | NP_082359.1 | 6 | 617 | ||
| GenBank mRNA | AK011243.1 | - | 6 | 618 | ||
| AK038378.1 | - | 6 | 587 | |||
| BC013532.1 | - | 5 | 527 | 4238801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chaf1b| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028083.4 | NP_082359.1 | 6 | 617 | ||
| GenBank mRNA | AK011243.1 | - | 6 | 618 | ||
| AK038378.1 | - | 6 | 587 | |||
| BC013532.1 | - | 5 | 527 | 4238801 | ||