siRNA Details
| Gene Symbol: NAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005967.3 | NP_005958.1 | 2 | 620 | ||
| XM_005268894.2 | XP_005268951.1 | 2 | 620 | |||
| GenBank mRNA | AK314229.1 | - | 2 | 400 | ||
| BC007756.2 | - | 2 | 354 | |||
| BC065931.1 | - | 2 | 407 | 6157017 | ||
| U48361.1 | - | 2 | 399 | |||
| X70991.1 | - | 1 | 95 | |||

NAB2
Genomic Map