siRNA Details
| Gene Symbol: Usp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130879.2 | NP_570949.2 | 28 | 5528 | ||
| GenBank mRNA | AK013309.1 | - | 1 | 1284 | ||
| BC058410.1 | - | 11 | 3174 | |||
| BC085481.1 | - | 1 | 1775 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp48| Gene Symbol: Usp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130879.2 | NP_570949.2 | 28 | 5528 | ||
| GenBank mRNA | AK013309.1 | - | 1 | 1284 | ||
| BC058410.1 | - | 11 | 3174 | |||
| BC085481.1 | - | 1 | 1775 | |||