siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030201.3 | NP_084477.1 | 4 | 712 | ||
| GenBank mRNA | AK030553.1 | - | 4 | 685 | ||
| AK034643.1 | - | 4 | 692 | |||
| AK049011.1 | - | 3 | 2193 | |||
| BC039701.1 | - | 3 | 653 | |||
| BC085181.1 | - | 4 | 672 | 30646642 | ||
| BC094508.1 | - | 4 | 655 | |||

Hspa13
Genomic Map