siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030201.3 | NP_084477.1 | 5 | 916 | ||
| GenBank mRNA | AK030553.1 | - | 6 | 985 | ||
| AK034643.1 | - | 5 | 896 | |||
| AK167839.1 | - | 5 | 859 | |||
| BC039701.1 | - | 4 | 857 | |||
| BC085181.1 | - | 5 | 876 | 30646642 | ||
| BC094508.1 | - | 6 | 955 | |||

Hspa13
Genomic Map