siRNA Details
| Gene Symbol: Adam33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163529.1 | NP_001157001.1 | 4, 5 | 643 | ||
| NM_033615.2 | NP_291093.2 | 4, 5 | 643 | |||
| GenBank mRNA | AB059632.1 | - | 4, 5 | 643 | ||
| AB059633.1 | - | 4, 5 | 643 | |||
| AF472524.1 | - | 4, 5 | 375 | |||
| AY054984.1 | - | 4, 5 | 343 | |||
| AY382193.1 | - | 4, 5 | 374 | |||
| AY382194.1 | - | 4, 5 | 374 | |||
| BC138139.1 | - | 4, 5 | 366 | 8861158 | ||

Adam33
Genomic Map