siRNA Details
| Gene Symbol: 1700102P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053216.2 | NP_444446.2 | 3, 4 | 533 | ||
| GenBank mRNA | AK007115.1 | - | 3, 4 | 533 | ||
| BC061048.1 | - | 3, 4 | 533 | 6770968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700102P08Rik| Gene Symbol: 1700102P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053216.2 | NP_444446.2 | 3, 4 | 533 | ||
| GenBank mRNA | AK007115.1 | - | 3, 4 | 533 | ||
| BC061048.1 | - | 3, 4 | 533 | 6770968 | ||