siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053246.3 | NP_444476.1 | 5, 6 | 735 | ||
| GenBank mRNA | AF418207.1 | - | 4, 5 | 619 | ||
| AK032352.1 | - | 5, 6 | 725 | |||
| AK043670.1 | - | 4, 5 | 647 | |||
| AK077612.1 | - | 5, 6 | 727 | |||
| AK139363.1 | - | 4, 5 | 623 | |||
| AK142621.1 | - | 4, 5 | 642 | |||
| AK156485.1 | - | 5, 6 | 735 | |||
| AK192791.1 | - | 219 | ||||
| BC004705.1 | - | 5, 6 | 696 | 3499419 | ||

Dok4
Genomic Map