siRNA Details
| Gene Symbol: Zmynd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053253.3 | NP_444483.2 | 2 | 332 | ||
| GenBank mRNA | AK132332.1 | - | 2 | 341 | ||
| BC052357.1 | - | 2 | 332 | 6772144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmynd10| Gene Symbol: Zmynd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053253.3 | NP_444483.2 | 2 | 332 | ||
| GenBank mRNA | AK132332.1 | - | 2 | 341 | ||
| BC052357.1 | - | 2 | 332 | 6772144 | ||