siRNA Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053262.3 | NP_444492.1 | 7 | 1263 | ||
| GenBank mRNA | AK033054.1 | - | 2 | 415 | ||
| AK154803.1 | - | 7 | 1280 | |||
| AK155174.1 | - | 7 | 1271 | |||
| AK155202.1 | - | 7 | 1275 | |||
| AK155327.1 | - | 7 | 1275 | |||
| AK158327.1 | - | 7 | 1268 | |||
| AK170939.1 | - | 7 | 1270 | |||
| BC038340.1 | - | 7 | 1257 | 4458725 | ||
| BC038665.1 | - | 1 | 71 | |||

Hsd17b11
Genomic Map