siRNA Details
| Gene Symbol: Ttyh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053273.2 | NP_444503.2 | 4 | 593 | ||
| GenBank mRNA | AF329682.1 | - | 4 | 536 | ||
| AK152172.1 | - | 4 | 545 | |||
| AK154835.1 | - | 4 | 601 | |||
| AK155166.1 | - | 4 | 594 | |||
| AK156393.1 | - | 4 | 546 | |||
| AK168408.1 | - | 4 | 546 | |||
| BC060740.1 | - | 4 | 536 | 4158813 | ||
| BY740853.1 | - | 4 | 573 | |||

Ttyh2
Genomic Map