siRNA Details
| Gene Symbol: Scgb3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289643.1 | NP_001276572.1 | 2, 3 | 446 | ||
| NM_001289644.1 | NP_001276573.1 | 2, 3 | 380 | |||
| GenBank mRNA | AF274959.1 | - | 2, 3 | 354 | ||
| AF274960.1 | - | 2, 3 | 420 | |||
| AF274961.1 | - | 2 | 684 | |||
| AF439546.1 | - | 2, 3 | 346 | |||
| AK136373.1 | - | 2, 3 | 358 | |||
| AK136396.1 | - | 2, 3 | 357 | |||
| BC061046.1 | - | 354 | 30301032 | |||

Scgb3a2
Genomic Map