siRNA Details
| Gene Symbol: Osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054049.2 | NP_473390.1 | 4 | 1463 | ||
| GenBank mRNA | AF370121.1 | - | 4 | 1463 | ||
| AK052147.1 | - | 4 | 1478 | |||
| AK161267.1 | - | 3 | 2516 | |||
| BC013504.1 | - | 4 | 1270 | 4224339 | ||

Osr2
Genomic Map