siRNA Details
| Gene Symbol: Pip4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054051.1 | NP_473392.1 | 6, 7 | 801 | ||
| GenBank mRNA | AB054987.1 | - | 2, 3 | 194 | ||
| AK089414.1 | - | 6, 7 | 791 | |||
| AK146559.1 | - | 6, 7 | 851 | |||
| AK170392.1 | - | 6, 7 | 728 | |||
| AK171497.1 | - | 6, 7 | 791 | |||
| AY050219.1 | - | 6, 7 | 630 | |||
| BC047282.1 | - | 6, 7 | 801 | 6491155 | ||

Pip4k2b
Genomic Map