siRNA Details
| Gene Symbol: Brdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079873.1 | NP_001073342.1 | 7 | 955 | ||
| NM_054054.2 | NP_473395.2 | 7 | 955 | |||
| GenBank mRNA | AB208640.1 | - | 5 | 648 | ||
| AF358660.1 | - | 7 | 990 | |||
| AK077137.1 | - | 7 | 975 | |||
| BY726053.1 | - | 3 | 281 | |||

Brdt
Genomic Map