siRNA Details
| Gene Symbol: Brdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054054.2 | NP_473395.2 | 10 | 1727 | ||
| GenBank mRNA | AF358660.1 | - | 10 | 1762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brdt| Gene Symbol: Brdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054054.2 | NP_473395.2 | 10 | 1727 | ||
| GenBank mRNA | AF358660.1 | - | 10 | 1762 | ||