siRNA Details
| Gene Symbol: Prosc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054057.4 | NP_473398.1 | 5, 6 | 511 | ||
| GenBank mRNA | AK032308.1 | - | 5, 6 | 461 | ||
| AK134549.1 | - | 5, 6 | 444 | |||
| AK160172.1 | - | 5, 6 | 465 | |||
| AK211128.1 | - | 1, 2 | 133 | |||
| AK213501.1 | - | 2, 3 | 149 | |||
| AK218333.1 | - | 2, 3 | 150 | |||

Prosc
Genomic Map