siRNA Details
| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054070.3 | NP_473411.2 | 6, 7 | 652 | ||
| GenBank mRNA | AF329695.1 | - | 6, 7 | 593 | ||
| AK159647.1 | - | 6, 7 | 616 | |||
| AK167964.1 | - | 6, 7 | 627 | |||
| AK168244.1 | - | 6, 7 | 626 | |||
| AK218541.1 | - | 2, 3 | 122 | |||
| BC056978.1 | - | 6, 7 | 611 | 6821746 | ||
| BC060064.1 | - | 6, 7 | 611 | |||

Afg3l1
Genomic Map