siRNA Details
| Gene Symbol: Mta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171052.1 | NP_001164523.1 | 15 | 1682 | ||
| NM_054082.2 | NP_473423.1 | 15 | 1679 | |||
| GenBank mRNA | AF288138.1 | - | 15 | 1633 | ||
| AK146721.1 | - | 15 | 1635 | |||
| AK146905.1 | - | 15 | 1635 | |||
| AK158866.1 | - | 14 | 1545 | |||

Mta3
Genomic Map