siRNA Details
| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054085.2 | NP_473426.2 | 5 | 1458 | ||
| GenBank mRNA | AF338872.1 | - | 5 | 1461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpk3| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054085.2 | NP_473426.2 | 5 | 1458 | ||
| GenBank mRNA | AF338872.1 | - | 5 | 1461 | ||