siRNA Details
| Gene Symbol: Tgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054089.4 | NP_473430.3 | 4 | 542 | ||
| GenBank mRNA | AF389908.1 | - | 4 | 411 | ||
| AK030953.1 | - | 4 | 534 | |||
| AK044895.1 | - | 4 | 543 | |||
| AK050044.1 | - | 4 | 472 | |||
| BC058183.1 | - | 4 | 546 | |||
| BC075728.1 | - | 4 | 523 | 6513964 | ||

Tgs1
Genomic Map