siRNA Details
| Gene Symbol: Olfr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054091.2 | NP_473432.2 | 1 | 928 | ||
| GenBank mRNA | AB061229.1 | - | 1 | 928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr74| Gene Symbol: Olfr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054091.2 | NP_473432.2 | 1 | 928 | ||
| GenBank mRNA | AB061229.1 | - | 1 | 928 | ||