siRNA Details
| Gene Symbol: CDC123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006023.2 | NP_006014.2 | 2, 3 | 438 | ||
| XM_005252638.3 | XP_005252695.1 | 2, 3 | 255 | |||
| GenBank mRNA | AK225634.1 | - | 2, 3 | 404 | ||
| AK289362.1 | - | 2, 3 | 175 | |||
| AK308790.1 | - | 2, 3 | 212 | |||
| BC001600.1 | - | 2, 3 | 188 | 3542611 | ||
| BC009598.1 | - | 2, 3 | 203 | 4134376 | ||
| BP367374.1 | - | 2, 3 | 438 | |||
| CR456966.1 | - | 2, 3 | 154 | |||
| D14878.1 | - | 2, 3 | 434 | |||
| U27112.1 | - | 2, 3 | 216 | |||

CDC123
Genomic Map