siRNA Details
| Gene Symbol: CPNE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280558.1 | NP_001267487.1 | 6 | 754 | ||
| NM_006032.3 | NP_006023.1 | 5 | 742 | |||
| XM_005268216.1 | XP_005268273.1 | 6 | 698 | |||
| XM_005268217.1 | XP_005268274.1 | 4 | 392 | |||
| GenBank mRNA | AB009288.1 | - | 5 | 542 | ||
| AK125195.1 | - | 4 | 1304 | |||
| AK222675.1 | - | 5 | 502 | |||
| AK293655.1 | - | 6 | 754 | |||
| AK294460.1 | - | 4 | 657 | |||
| AK314182.1 | - | 5 | 555 | |||
| AL137688.1 | - | 3 | 1953 | |||
| BC018046.1 | - | 5 | 548 | 4799650 | ||

CPNE6
Genomic Map