siRNA Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081977.2 | NP_001075446.1 | 10 | 1861 | ||
| NM_080563.4 | NP_542130.1 | 9 | 1787 | |||
| GenBank mRNA | AK038586.1 | - | 9 | 1679 | ||
| AK133544.1 | - | 7 | 1401 | |||
| AK172904.1 | - | 8 | 1623 | |||
| BC030187.1 | - | 9 | 1752 | 5345434 | ||

Rnf144a
Genomic Map