siRNA Details
| Gene Symbol: Timp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080639.3 | NP_542370.3 | 5 | 727 | ||
| GenBank mRNA | AF282730.1 | - | 5 | 658 | ||
| AK038820.1 | - | 5 | 726 | |||
| AK079347.1 | - | 5 | 728 | |||
| AK140367.1 | - | 5 | 727 | |||
| BC064046.1 | - | 5 | 707 | 875422 | ||

Timp4
Genomic Map