siRNA Details
| Gene Symbol: SPATA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135773.1 | NP_001129245.1 | 2 | 676 | ||
| NM_006038.3 | NP_006029.1 | 2 | 592 | |||
| XM_006723894.1 | XP_006723957.1 | 2 | 512 | |||
| XM_011529116.1 | XP_011527418.1 | 2 | 549 | |||
| GenBank mRNA | AB018300.1 | - | 2 | 510 | ||
| AK023229.1 | - | 2 | 547 | |||
| AK289644.1 | - | 2 | 676 | |||
| BC009481.2 | - | 2 | 499 | 3953522 | ||
| CB992178.1 | - | 2 | 607 | |||
| U28164.1 | - | 2 | 557 | |||

SPATA2
Genomic Map