siRNA Details
| Gene Symbol: Bmp2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080708.1 | NP_542439.1 | 2 | 497 | ||
| GenBank mRNA | AK046752.1 | - | 2 | 462 | ||
| AY050249.1 | - | 2 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp2k| Gene Symbol: Bmp2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080708.1 | NP_542439.1 | 2 | 497 | ||
| GenBank mRNA | AK046752.1 | - | 2 | 462 | ||
| AY050249.1 | - | 2 | 497 | |||