siRNA Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080849.3 | NP_543125.1 | 15 | 2273 | ||
| GenBank mRNA | AF407579.1 | - | 15 | 2223 | ||
| AK014546.1 | - | 13 | 2449 | |||
| AK154358.1 | - | 15 | 2236 | |||
| BC057995.1 | - | 14 | 2355 | |||
| BC070457.1 | - | 15 | 2222 | 30603080 | ||

Nek8
Genomic Map