siRNA Details
| Gene Symbol: Zcchc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080855.2 | NP_543131.1 | 5, 6 | 587 | ||
| GenBank mRNA | AB030244.1 | - | 5, 6 | 598 | ||
| BC132449.1 | - | 5, 6 | 585 | 40130726 | ||
| BC137853.1 | - | 5, 6 | 585 | 8860872 | ||
| CB518791.1 | - | 3, 4 | 233 | |||

Zcchc14
Genomic Map