siRNA Details
| Gene Symbol: Zcchc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080855.2 | NP_543131.1 | 13 | 4329 | ||
| GenBank mRNA | AB030244.1 | - | 13 | 4339 | ||
| AK035149.1 | - | 1 | 1075 | |||
| AK041107.1 | - | 1 | 2838 | |||
| AK122315.1 | - | 1 | 3412 | |||
| BC009020.1 | - | 2 | 2107 | |||
| BC094454.1 | - | 2 | 1580 | |||
| BC132449.1 | - | 13 | 4328 | 40130726 | ||
| BC137853.1 | - | 13 | 4329 | 8860872 | ||

Zcchc14
Genomic Map