siRNA Details
| Gene Symbol: UBE4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105562.2 | NP_001099032.1 | 4 | 1053 | ||
| NM_006048.4 | NP_006039.2 | 4 | 1053 | |||
| XM_005263422.1 | XP_005263479.1 | 4 | 877 | |||
| XM_011540488.2 | XP_011538790.1 | 3 | 176 | |||
| XM_011540489.2 | XP_011538791.1 | 4 | 877 | |||
| XM_011540490.1 | XP_011538792.1 | 4 | 877 | |||
| XM_017000036.1 | XP_016855525.1 | 5 | 717 | |||
| GenBank mRNA | AB014584.3 | - | 4 | 944 | ||
| AB028839.1 | - | 4 | 1061 | |||
| AF043117.1 | - | 4 | 450 | |||
| AF331520.1 | - | 4 | 412 | |||
| AK292444.1 | - | 4 | 535 | |||
| AK304484.1 | - | 2 | 40 | |||
| BC093696.1 | - | 4 | 443 | 7939541 | ||
| BC143458.1 | - | 4 | 443 | |||
| BC143460.1 | - | 4 | 443 | |||
| JF289274.1 | - | 4 | 365 | |||

UBE4B
Genomic Map