siRNA Details
| Gene Symbol: Cntnap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130457.2 | NP_569724.2 | 4 | 564 | ||
| GenBank mRNA | AF333770.1 | - | 4 | 529 | ||
| AK042607.1 | - | 4 | 553 | |||
| AK053611.1 | - | 4 | 556 | |||
| AK081891.1 | - | 4 | 557 | |||
| AK142816.1 | - | 4 | 560 | |||
| AK173255.1 | - | 4 | 539 | |||
| BC034628.1 | - | 4 | 564 | 5369648 | ||

Cntnap4
Genomic Map