siRNA Details
Gene Symbol: Olfr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168503.1 | NP_001161975.1 | 3 | 2261 | ||
NM_130866.4 | NP_570936.1 | 4 | 3130 | |||
GenBank mRNA | AK028467.1 | - | 4 | 3054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168503.1 | NP_001161975.1 | 3 | 2261 | ||
NM_130866.4 | NP_570936.1 | 4 | 3130 | |||
GenBank mRNA | AK028467.1 | - | 4 | 3054 |