siRNA Details
| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130880.1 | NP_570950.1 | 5 | 564 | ||
| GenBank mRNA | AJ430384.1 | - | 5 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud7a| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130880.1 | NP_570950.1 | 5 | 564 | ||
| GenBank mRNA | AJ430384.1 | - | 5 | 564 | ||